Tantalum ≥99.95% (metals basis), foil, annealed, Thickness: 0.127 mm (0.005 in)
Supplier: Thermo Fisher Scientific
10352.FI 10352.GH 10352.GW
10352.FIEA 89.2 EUR
10352.FI 10352.GH 10352.GW
Tantalum ≥99.95% (metals basis), foil, annealed, Thickness: 0.127 mm (0.005 in)
Tantalum
Purity: 99.95% CAS No.: 7440-25-7 Physical Form: ≈5.3g/50x50mm, Temper: soft Odour: Odourless MDL No.: MFCD00011252
Formula: Ta
MW: 180.95 g/mol
Boiling Pt: 5427
Melting Pt: 2996
Density: 16.69 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011252
CAS Number: 7440-25-7
EINECS: 231-135-5
Merck Index: 12,09223