Tantalum ≥99.95% (metals basis), thin foil, Thickness: 0.0127 mm (0.0005 in)
Supplier: Thermo Fisher Scientific
14266.8S 14266.GD 14266.FY
14266.8SEA 1030 EUR
14266.8S 14266.GD 14266.FY
Tantalum ≥99.95% (metals basis), thin foil, Thickness: 0.0127 mm (0.0005 in)
Tantalum
Purity: 99.95% CAS No.: 7440-25-7 Physical Form: ≈2.1g/100x100mm, certified pinhole free Odour: Odourless MDL No.: MFCD00011252
Formula: Ta
MW: 180.95 g/mol
Boiling Pt: 5427
Melting Pt: 2996
Density: 16.69 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011252
CAS Number: 7440-25-7
EINECS: 231-135-5
Merck Index: 12,09223