Tantalum, gauze woven from wire 30 mesh woven from 0.076mm (0.003in)
Supplier: Thermo Fisher Scientific
43965.GN 43965.GY
43965.GNEA 336 EUR
43965.GN 43965.GY
Tantalum, gauze woven from wire 30 mesh woven from 0.076mm (0.003in)
Tantalum
Formula: Ta
MW: 180.95 g/mol
Boiling Pt: 5427
Melting Pt: 2996
Density: 16.69 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011252
CAS Number: 7440-25-7
EINECS: 231-135-5
Merck Index: 12,09223