Tantalum (50 ppm Nb) ≥99.98% (metals basis), powder, Puratronic® -100 mesh
Supplier: Thermo Fisher Scientific
Warning
10344.14 10344.22 10344.88
10344.14EA 179 EUR
10344.14 10344.22 10344.88
Tantalum (50 ppm Nb) ≥99.98% (metals basis), powder, Puratronic® -100 mesh
Tantalum
Purity: 99.98% CAS No.: 7440-25-7 Appearance: Black Odour: Odourless UN No.: 3089 MDL No.: MFCD00011252
Formula: Ta
MW: 180.95 g/mol
Boiling Pt: 5427
Melting Pt: 2996
Density: 16.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011252
CAS Number: 7440-25-7
EINECS: 231-135-5
UN: 3089
ADR: 4.1 III
Merck Index: 12,09223