Vanadium ≥99.5% (metals basis), rods, Ø 6.35 mm (0.25 in)
Supplier: Thermo Fisher Scientific
13996.BQ 13996.BU 13996.BY
13996.BQEA 166 EUR
13996.BQ 13996.BU 13996.BY
Vanadium ≥99.5% (metals basis), rods, Ø 6.35 mm (0.25 in)
Vanadium
Purity: 99.5% CAS No.: 7440-62-2 Physical Form: ≈1.93g/cm Odour: Odourless MDL No.: MFCD00011453
Formula: V
MW: 50.94 g/mol
Boiling Pt: 3477
Melting Pt: 1896
Density: 6.015 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011453
CAS Number: 7440-62-2
EINECS: 231-171-1
Merck Index: 12,10053