Vanadium ≥99.8% (metals basis), wire, Ø 1.5 mm (0.06 in)
Supplier: Thermo Fisher Scientific
42224.BS 42224.BW 42224.CB
42224.BSEA 70.8 EUR
42224.BS 42224.BW 42224.CB
Vanadium ≥99.8% (metals basis), wire, Ø 1.5 mm (0.06 in)
Vanadium
Purity: 99.8% CAS No.: 7440-62-2 Physical Form: ≈10.8g/m Odour: Odourless MDL No.: MFCD00011453
Formula: V
MW: 50.94 g/mol
Boiling Pt: 3477
Melting Pt: 1896
Density: 6.015 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011453
CAS Number: 7440-62-2
EINECS: 231-171-1
Merck Index: 12,10053