Vanadium ≥99.8% (metals basis), foil, Thickness: 0.075 mm (0.003 in)
Supplier: Thermo Fisher Scientific
44106.FY 44106.GK
44106.FYEA 200 EUR
44106.FY 44106.GK
Vanadium ≥99.8% (metals basis), foil, Thickness: 0.075 mm (0.003 in)
Vanadium
Purity: 99.8% CAS No.: 7440-62-2 Physical Form: ≈2.29g/50x100mm Odour: Odourless MDL No.: MFCD00011453
Formula: V
MW: 50.94 g/mol
Boiling Pt: 3477
Melting Pt: 1896
Density: 6.015 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011453
CAS Number: 7440-62-2
EINECS: 231-171-1
Merck Index: 12,10053