Chromium, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
374849-250G 374849-50G
SIAL374849-250GEA 474 EUR
SIAL374849-250G SIAL374849-50G
Chromium, Sigma-Aldrich®
Chromium
Formula: Cr
MW: 52 g/mol
Boiling Pt: 2469
Melting Pt: 1840
Density: 7.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010944
CAS Number: 7440-47-3
EINECS: 231-157-5
Merck Index: 12,02288