Molybdenum ≥99.95% (metals basis), rods, Ø 12.7 mm (0.5 in)
Supplier: Thermo Fisher Scientific
10034.18 10034.30
10034.18EA 110 EUR
10034.18 10034.30
Molybdenum ≥99.95% (metals basis), rods, Ø 12.7 mm (0.5 in)
Molybdenum
Purity: 99.95% CAS No.: 7439-98-7 Physical Form: ≈3.8cm/50g MDL No.: MFCD00003465
Formula: Mo
MW: 95.94 g/mol
Boiling Pt: 3617
Melting Pt: 2622
Density: 10.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
Merck Index: 12,06317