Molybdenum ≥99.94% (metals basis), wire, annealed, Ø 1.0 mm (0.04 in)
Supplier: Thermo Fisher Scientific
10039.G5 10039.G9 10039.H4
10039.G5EA 214 EUR
10039.G5 10039.G9 10039.H4
Molybdenum ≥99.94% (metals basis), wire, annealed, Ø 1.0 mm (0.04 in)
Molybdenum
Purity: 99.94% CAS No.: 7439-98-7 Physical Form: ≈8.0g/m, Temper: soft MDL No.: MFCD00003465
Formula: Mo
MW: 95.94 g/mol
Boiling Pt: 3617
Melting Pt: 2622
Density: 10.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
Merck Index: 12,06317