Molybdenum, powder, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

510092-250G 510092-50G
SIAL510092-250GEA 179 EUR
SIAL510092-250G SIAL510092-50G
Molybdenum, powder, Sigma-Aldrich®
Molybdenum
Formula: Mo
Boiling Pt: 5560 °C (1013 hPa)
Melting Pt: 2622 °C
Density: 10.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
UN: 3089
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR