Tungsten ≥99.9% (metals basis), slug, Length: 6.4 mm (0.25 in), Ø 17.5 mm (0.689 in)
Supplier: Thermo Fisher Scientific
00525.KG 00525.KI
00525.KGEA 152 EUR
00525.KG 00525.KI
Tungsten ≥99.9% (metals basis), slug, Length: 6.4 mm (0.25 in), Ø 17.5 mm (0.689 in)
Tungsten
Purity: 99.9% CAS No.: 7440-33-7 Physical Form: ≈30g/slug MDL No.: MFCD00011461
Formula: W
Boiling Pt: 5660 °C (1013 hPa)
Melting Pt: 3410 °C
Density: 19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
Merck Index: 12,09945