Tungsten ≥99.95% (metals basis), rods, annealed, Ø 6.3 mm (0.25 in)
Supplier: Thermo Fisher Scientific
10406.BS 10406.BW
10406.BSEA 149 EUR
10406.BS 10406.BW
Tungsten ≥99.95% (metals basis), rods, annealed, Ø 6.3 mm (0.25 in)
Tungsten
Purity: 99.95% CAS No.: 7440-33-7 Physical Form: ≈6.01g/cm, Temper: soft MDL No.: MFCD00011461
Formula: W
MW: 183.84 g/mol
Boiling Pt: 5660
Melting Pt: 3410
Density: 19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
Merck Index: 12,09945