Tungsten ≥99.95% (metals basis), foil, Thickness: 0.25 mm (0.01 in)
Supplier: Thermo Fisher Scientific
10415.FI 10415.GH 10415.HP
10415.FIEA 110 EUR
10415.FI 10415.GH 10415.HP
Tungsten ≥99.95% (metals basis), foil, Thickness: 0.25 mm (0.01 in)
Tungsten
Purity: 99.95% CAS No.: 7440-33-7 Physical Form: ≈12g/50x50mm MDL No.: MFCD00011461
Formula: W
Boiling Pt: 5660 °C (1013 hPa)
Melting Pt: 3410 °C
Density: 19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
Merck Index: 12,09945