Tungsten, gauze woven from wire 100 mesh, Ø 0.0509 mm (0.002 in)
Supplier: Thermo Fisher Scientific
40925.FL 40925.GT 40925.HB
40925.FLEA 419 EUR
40925.FL 40925.GT 40925.HB
Tungsten, gauze woven from wire 100 mesh, Ø 0.0509 mm (0.002 in)
Tungsten
CAS No.: 7440-33-7 Physical Form: Wire Cloth MDL No.: MFCD00011461
Formula: W
MW: 183.84 g/mol
Boiling Pt: 5660
Melting Pt: 3410
Density: 19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
Merck Index: 12,09945