Tungsten ≥99.9% (metals basis), powder APS 1-5 µm
Supplier: Thermo Fisher Scientific
Warning
10400.09 10400.22 10400.36 10400.A3
10400.09EA 62.1 EUR
10400.09 10400.22 10400.36 10400.A3
Tungsten ≥99.9% (metals basis), powder APS 1-5 µm
Tungsten
Purity: 99.9% CAS No.: 7440-33-7 Physical Form: Powder Appearance: Grey UN No.: 3089 MDL No.: MFCD00011461
Formula: W
MW: 183.84 g/mol
Boiling Pt: 5660
Melting Pt: 3410
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
UN: 3089
ADR: 4.1 III
Merck Index: 12,09945