Tungsten ≥99.95% (metals basis), powder -200+325 mesh
Supplier: Thermo Fisher Scientific
Warning
42477.22 42477.36 42477.A3
42477.22EA 77.5 EUR
42477.22 42477.36 42477.A3
Tungsten ≥99.95% (metals basis), powder -200+325 mesh
Tungsten
Purity: 99.95% CAS No.: 7440-33-7 Appearance: Grey UN No.: 3089 MDL No.: MFCD00011461
Formula: W
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
UN: 3089
ADR: 4.1,III