Rhenium, Sigma-Aldrich®
Supplier: Merck
MQ100
267317-13.2G
267317-3.3G
SIAL267317-13.2GEA
1780
EUR
SIAL267317-13.2G
SIAL267317-3.3G
Rhenium, Sigma-Aldrich®
Rhenium
Formula:
Re MW: 186.21 g/mol Boiling Pt: 5630 Melting Pt: 3180 Density: 21.02 g/cm³ (20 °C) |
MDL Number:
MFCD00011195 CAS Number: 7440-15-5 EINECS: 231-124-5 Merck Index: 12,08343 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...