Cobalt ≥99.95% (metals basis), foil, Thickness: 0.05 mm (0.002 in)
Supplier: Thermo Fisher Scientific
Danger
42657.FF 42657.FI 42657.GH
42657.FFEA 98 EUR
42657.FF 42657.FI 42657.GH
Cobalt ≥99.95% (metals basis), foil, Thickness: 0.05 mm (0.002 in)
Cobalt
Purity: 99.95% CAS No.: 7440-48-4 Physical Form: ≈0.28g/25x25mm Odour: Odourless MDL No.: MFCD00010935
Formula: Co
MW: 58.93 g/mol
Boiling Pt: 2870
Melting Pt: 1495
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
Merck Index: 12,02488