Cobalt ≥99.5% (metals basis), powder -400 mesh
Supplier: Thermo Fisher Scientific
Danger
44085.30 44085.A1
44085.30EA 442 EUR
44085.30 44085.A1
Cobalt ≥99.5% (metals basis), powder -400 mesh
Cobalt
Purity: 99.5% CAS No.: 7440-48-4 Appearance: Black Odour: Odourless UN No.: 3089 MDL No.: MFCD00010935
Formula: Co
MW: 58.93 g/mol
Boiling Pt: 2870
Melting Pt: 1495
Storage Temperature: Ambient
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
UN: 3089
ADR: 4.1 II
Merck Index: 12,02488