Cobalt, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
203076-25G 203076-5G
SIAL203076-25GEA 506 EUR
SIAL203076-25G SIAL203076-5G
Cobalt, Sigma-Aldrich®
Cobalt
Formula: Co
MW: 58.93 g/mol
Boiling Pt: 2870
Melting Pt: 1495
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
Merck Index: 12,02488