Iron ≥99.99% (metals basis), wire, Ø 0.5 mm (0.02 in)
Supplier: Thermo Fisher Scientific
40498.G1 40498.G5
40498.G1EA 152 EUR
40498.G1 40498.G5
Iron ≥99.99% (metals basis), wire, Ø 0.5 mm (0.02 in)
Iron
Purity: 99.99% CAS No.: 7439-89-6 Physical Form: ≈1.5g/m MDL No.: MFCD00010999
Formula: Fe
Boiling Pt: 3000 °C (1013 hPa)
Melting Pt: 1535 °C
Density: 7.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010999
CAS Number: 7439-89-6
EINECS: 231-096-4
Merck Index: 13,05109