Iron ≥99.99% (metals basis), pieces, irregularly shaped 3.2-6.4 mm (0.125-0.25 in)
Supplier: Thermo Fisher Scientific
45090.A1 45090.36 45090.22
45090.A1EA 407 EUR
45090.A1 45090.36 45090.22
Iron ≥99.99% (metals basis), pieces, irregularly shaped 3.2-6.4 mm (0.125-0.25 in)
Iron
Purity: 99.99% CAS No.: 7439-89-6 Appearance: Grey MDL No.: MFCD00010999
Formula: Fe
Boiling Pt: 3000 °C (1013 hPa)
Melting Pt: 1535 °C
Density: 7.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010999
CAS Number: 7439-89-6
EINECS: 231-096-4
Merck Index: 13,05109