Iridium ≥99.8% (metals basis), wire, Ø 0.25 mm (0.01 in)
Supplier: Thermo Fisher Scientific
11432.BSEA 275 EUR
11432.BS 11432.BW 11432.CA
Iridium ≥99.8% (metals basis), wire, Ø 0.25 mm (0.01 in)
Iridium
Purity: 99.8% CAS No.: 7439-88-5 Physical Form: ≈1.11g/m MDL No.: MFCD00011062
Formula: Ir
MW: 192.22 g/mol
Boiling Pt: 5300
Melting Pt: 2450
Storage Temperature: Ambient
MDL Number: MFCD00011062
CAS Number: 7439-88-5
EINECS: 231-095-9