Nickel ≥99.7% (metals basis), wire, Ø 0.63 mm (0.025 in)
Supplier: Thermo Fisher Scientific
Danger
00223.G9 00223.H4
00223.G9EA 143 EUR
00223.G9 00223.H4
Nickel ≥99.7% (metals basis), wire, Ø 0.63 mm (0.025 in)
Nickel
Purity: 99.7% CAS No.: 7440-02-0 Physical Form: ≈2.8g/m Odour: Odourless MDL No.: MFCD00011137
Formula: Ni
Boiling Pt: 3007 °C (1013 hPa)
Melting Pt: 1453 °C
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1,III
Merck Index: 12,06582