Nickel ≥99.5% (metals basis), foil, annealed, Thickness: 0.025 mm (0.001 in)
Supplier: Thermo Fisher Scientific
Danger
00228.FT 00228.FW
00228.FTEA 107 EUR
00228.FT 00228.FW
Nickel ≥99.5% (metals basis), foil, annealed, Thickness: 0.025 mm (0.001 in)
Nickel
Purity: 99.5% CAS No.: 7440-02-0 Physical Form: ≈1.4g/25x250mm Odour: Odourless MDL No.: MFCD00011137
Formula: Ni
Boiling Pt: 3007 °C (1013 hPa)
Melting Pt: 1453 °C
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1,III
Merck Index: 12,06582