Nickel ≥99.8% (metals basis), spherical powder -300 mesh
Supplier: Thermo Fisher Scientific
Danger
10581.09 10581.22 10581.36 10581.A3
10581.09EA 32.6 EUR
10581.09 10581.22 10581.36 10581.A3
Nickel ≥99.8% (metals basis), spherical powder -300 mesh
Nickel
Purity: 99.8% CAS No.: 7440-02-0 Odour: Odourless UN No.: 3089 MDL No.: MFCD00011137
Formula: Ni
Boiling Pt: 2371 °C (1013 hPa)
Melting Pt: 1455 °C
Density: 8.9 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1,II