Nickel ≥99.994% (metals basis), foil, Puratronic®, Thickness: 0.5 mm (0.02 in)
Supplier: Thermo Fisher Scientific
Danger
12044.FF 12044.FI 12044.GH
12044.FFEA 153 EUR
12044.FF 12044.FI 12044.GH
Nickel ≥99.994% (metals basis), foil, Puratronic®, Thickness: 0.5 mm (0.02 in)
Nickel
Purity: 99.994% CAS No.: 7440-02-0 Physical Form: ≈2.8g/25x25mm Odour: Odourless MDL No.: MFCD00011137
Formula: Ni
MW: 58.69 g/mol
Boiling Pt: 3007
Melting Pt: 1453
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1 III
Merck Index: 12,06582