Nickel ≥99% (metals basis), foil, annealed, Thickness: 0.025 mm (0.001 in)
Supplier: Thermo Fisher Scientific
Danger
12722.CH 12722.CJ 12722.RX
12722.CHEA 72.4 EUR
12722.CH 12722.CJ 12722.RX
Nickel ≥99% (metals basis), foil, annealed, Thickness: 0.025 mm (0.001 in)
Nickel
Purity: 99% CAS No.: 7440-02-0 Physical Form: ≈13.6g/20x30cm, Temper: soft Odour: Odourless MDL No.: MFCD00011137
Formula: Ni
MW: 58.69 g/mol
Boiling Pt: 3007
Melting Pt: 1453
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1 III
Merck Index: 12,06582