Nickel, gauze woven from wire 50 mesh, Ø 0.05 mm (0.002 in)
Supplier: Thermo Fisher Scientific
Danger
40914.FL 40914.GT 40914.HB
40914.FLEA 63.8 EUR
40914.FL 40914.GT 40914.HB
Nickel, gauze woven from wire 50 mesh, Ø 0.05 mm (0.002 in)
Nickel
CAS No.: 7440-02-0 Physical Form: Wire Cloth Odour: Odourless MDL No.: MFCD00011137
Formula: Ni
MW: 58.69 g/mol
Boiling Pt: 3007
Melting Pt: 1453
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1 III
Merck Index: 12,06582