Nickel ≥99.98% (metals basis), slug, Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)
Supplier: Thermo Fisher Scientific
Danger
42326.09 42326.18 42326.30
42326.09EA 52.5 EUR
42326.09 42326.18 42326.30
Nickel ≥99.98% (metals basis), slug, Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)
Nickel
Purity: 99.98% CAS No.: 7440-02-0 Physical Form: ≈0.45g/slug Odour: Odourless MDL No.: MFCD00011137
Formula: Ni
MW: 58.69 g/mol
Boiling Pt: 3007
Melting Pt: 1453
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1 III
Merck Index: 12,06582