Nickel ≥99.98% (metals basis), wire, annealed, Ø 0.5 mm (0.02 in)
Supplier: Thermo Fisher Scientific
Danger
43132.H4 43132.G9 43132.G5
43132.H4EA 681 EUR
43132.H4 43132.G9 43132.G5
Nickel ≥99.98% (metals basis), wire, annealed, Ø 0.5 mm (0.02 in)
Nickel
Purity: 99.98% CAS No.: 7440-02-0 Physical Form: ≈1.75g/m, Temper: soft Odour: Odourless MDL No.: MFCD00011137
Formula: Ni
MW: 58.69 g/mol
Boiling Pt: 3007
Melting Pt: 1453
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1 III
Merck Index: 12,06582