Palladium ≥99.95% (metals basis), rods, Ø 6.35 mm
Supplier: Thermo Fisher Scientific
12557.DH 12557.DM
12557.DHEA 0 EUR
12557.DH 12557.DM
Palladium ≥99.95% (metals basis), rods, Ø 6.35 mm
Palladium
Purity: 99.95% CAS No.: 7440-05-3 Physical Form: ≈3.80g/cm Odour: Odourless MDL No.: MFCD00011167
Formula: Pd
Boiling Pt: 3167 °C (1013 hPa)
Melting Pt: 1555 °C
Density: 12.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
Merck Index: 12,07121