Palladium 99.99% (trace metals basis), powder - 20 mesh
Supplier: Thermo Fisher Scientific
Warning
193690010 193690050 193690250
ACRO193690010EA 338 EUR
ACRO193690010 ACRO193690050 ACRO193690250
Palladium 99.99% (trace metals basis), powder - 20 mesh
Palladium
Formula: Pd
Melting Pt: 1555 °C
Density: 12,02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
UN: 3089
ADR: 4.1,II
Order Now
Specification Test Results
Appearance Grey to black Powder
Total trace metal impurities ≤0.01 %