Palladium ≥99.999% (trace metals basis), powder, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
203939-1G 203939-5G
SIAL203939-1GEA 344 EUR
SIAL203939-1G SIAL203939-5G
Palladium ≥99.999% (trace metals basis), powder, Sigma-Aldrich®
Palladium
Formula: Pd
Density: 12.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
ADR: 4.1,II