Platinum ≥99.9% (metals basis), gauze woven from wire 52 mesh, Ø 0.1 mm (0.004 in)
Supplier: Thermo Fisher Scientific
Synonyms: Platinium
10283.FF 10283.FI 10283.GH 10283.FL
10283.FFEA 301 EUR
10283.FF 10283.FI 10283.GH 10283.FL
Platinum ≥99.9% (metals basis), gauze woven from wire 52 mesh, Ø 0.1 mm (0.004 in)
Platinum
Purity: 99.9% CAS No.: 7440-06-4 Physical Form: ≈0.47g/25x25mm, Wire Cloth Odour: Odourless MDL No.: MFCD00011179
Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827
Melting Pt: 1769
Density: 21.447 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689