Platinum ≥99.997% (metals basis), wire, Premion®, Ø 0.1 mm (0.004 in)
Supplier: Thermo Fisher Scientific
Synonyms: Platinium
10959.G1 10959.G5
10959.G1EA 173 EUR
10959.G1 10959.G5
Platinum ≥99.997% (metals basis), wire, Premion®, Ø 0.1 mm (0.004 in)
Platinum
Purity: 99.997% CAS No.: 7440-06-4 Physical Form: ≈0.17g/m Odour: Odourless MDL No.: MFCD00011179
Formula: Pt
Boiling Pt: 3827 °C (1013 hPa)
Melting Pt: 1769 °C
Density: 21.447 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689