Platinum ≥99.98% (metals basis), sponge -60 mesh
Supplier: Thermo Fisher Scientific
Synonyms: Platinium
12074.03 12074.06
12074.03EA 241 EUR
12074.03 12074.06
Platinum ≥99.98% (metals basis), sponge -60 mesh
Platinum
Purity: 99.98% CAS No.: 7440-06-4 Odour: Odourless MDL No.: MFCD00011179
Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827
Melting Pt: 1769
Density: 21.447 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689