Platinum ≥99.98% (metals basis), powder -200 mesh
Supplier: Thermo Fisher Scientific
12075.03 12075.06 12075.77
12075.03EA 321 EUR
12075.03 12075.06 12075.77
Platinum ≥99.98% (metals basis), powder -200 mesh
Platinum
Purity: 99.98% CAS No.: 7440-06-4 Odour: Odourless MDL No.: MFCD00011179
Formula: Pt
Boiling Pt: 4530 °C (1013 hPa)
Melting Pt: 1773 °C
Density: 21.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
UN: 3089
ADR: 4.1,II