Platinum ≥99.95% (metals basis), granules ≤ 6.35 mm (0.25 in)
Supplier: Thermo Fisher Scientific
Synonyms: Platinium
12236.03 12236.06 12236.14
12236.03EA 344 EUR
12236.03 12236.06 12236.14
Platinum ≥99.95% (metals basis), granules ≤ 6.35 mm (0.25 in)
Platinum
Purity: 99.95% CAS No.: 7440-06-4 Odour: Odourless MDL No.: MFCD00011179
Formula: Pt
Boiling Pt: 3827 °C (1013 hPa)
Melting Pt: 1769 °C
Density: 21.447 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689