Platinum 99,95 Metals basis, wire, hard, Ø 0.0508 mm (0.002 in)
Supplier: Thermo Fisher Scientific
Synonyms: Platinium
13479.G1 13479.G5 13479.G9
13479.G1EA 67.9 EUR
13479.G1 13479.G5 13479.G9
Platinum 99,95 Metals basis, wire, hard, Ø 0.0508 mm (0.002 in)
Platinum
Purity: 99.95% CAS No.: 7440-06-4 Physical Form: ≈0.043g/m, Temper: as drawn Odour: Odourless MDL No.: MFCD00011179
Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827
Melting Pt: 1769
Density: 21.447 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689