Rhodium ≥99.95% (metals basis), powder -325 mesh
Supplier: Thermo Fisher Scientific
Warning
38514.02 38514.04
38514.02EA 1380 EUR
38514.02 38514.04
Rhodium ≥99.95% (metals basis), powder -325 mesh
Rhodium
Purity: 99.95% CAS No.: 7440-16-6 Odour: Odourless UN No.: 3089 MDL No.: MFCD00011201
Formula: Rh
Storage Temperature: Ambient
MDL Number: MFCD00011201
CAS Number: 7440-16-6
EINECS: 231-125-0
UN: 3089
ADR: 4.1,III