Soda lime ACS, with indicator
Supplier: Thermo Fisher Scientific
|
Danger
|
44697.36
44697.A4
44697.36EA
78.1
EUR
44697.36
44697.A4
Soda lime ACS, with indicator
Soda lime
CAS No.: 8006-28-8
Physical Form: 4-8 Mesh
Sensitivity: Air Sensitive and Hygroscopic
Odour: Odourless
Merck Reference: 14,8569
UN No.: 1907
MDL No.: MFCD00134094
Storage Temperature:
Ambient |
MDL Number:
MFCD00134094 CAS Number: 8006-28-8 UN: 1907 ADR: 8,III Merck Index: 14,8569 |