2-Methyl-2-pentenal, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
W319406-1KG-K
W319406-4KG-K
SIALW319406-1KG-KEA
219
EUR
SIALW319406-1KG-K
SIALW319406-4KG-K
2-Methyl-2-pentenal, Sigma-Aldrich®
2-Methyl-2-pentenal
Formula:
C₆H₁₀O Boiling Pt: 135…138 °C (1013 hPa) Density: 0.855 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006978 CAS Number: 623-36-9 EINECS: 210-789-5 UN: 1988 ADR: 3,III |