4-Propenylveratrole, Sigma-Aldrich®

Supplier: Merck

Synonyms: 4-(1-Propenyl)pyrocatechol dimethyl ether, 4-(1-Propenyl)-1,2-dimethoxybenzene, 1,2-Dimethoxy-4-Propenylbenzene, 1,2-Dimethoxy-4-(1-propenyl)benzene, Methyl isoeugenol, O-Methyl Isoeugenol

M-Clarity™ quality level = MQ300

W247618-10KG-K W247618-1KG-K W247618-250G-K
SIALW247618-10KG-KEA 1200 EUR
SIALW247618-10KG-K SIALW247618-1KG-K SIALW247618-250G-K
4-Propenylveratrole, Sigma-Aldrich®
4-Propenylveratrole
Formula: C₁₁H₁₄O₂
Density: 1.06 g/cm³ (20 °C)
MDL Number: MFCD00009282
CAS Number: 93-16-3
EINECS: 202-224-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR