Paraffin wax tested according to Ph Eur, high viscosity, Sigma-Aldrich®
Supplier: Merck
Synonyms: Wax Paraffin, Paraffin wax
M-Clarity™ quality level = MQ100
76234-10L 76234-1L
SIAL76234-10LEA 400 EUR
SIAL76234-10L SIAL76234-1L
Paraffin wax tested according to Ph Eur, high viscosity, Sigma-Aldrich®
Paraffin
Density: 0.86 g/cm³ (20 °C)
MDL Number: MFCD00132833
CAS Number: 8002-74-2
EINECS: 232-384-2
Merck Index: 13,07263