Blue Dextran stain, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

D5751-5G D5751-1G
SIALD5751-5GEA 331 EUR
SIALD5751-5G SIALD5751-1G
Blue Dextran stain, Sigma-Aldrich®
Blue Dextran

Blue dextran are used in affinity chromatography, gel filtration chromatography, protein chromatography and molecular weight markers.


  • Void volume marker for gel filtration columns
  • Prepared from dextran with an average mol. wt. of 2,000,000


Blue dextran has been used to study Alzheimer′s disease.

Formula: (C₂₉H₂₁N₇O₁₂S₃·3Na)n
MDL Number: MFCD00130661
CAS Number: 87915-38-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR