Beeswax, Sigma-Aldrich®
Supplier: Merck
Synonyms: Wax, bees
M-Clarity™ quality level = MQ100
243248-100G 243248-500G
SIAL243248-100GEA 59.3 EUR
SIAL243248-100G SIAL243248-500G
Beeswax, Sigma-Aldrich®
Beeswax
Melting Pt: ∼64 °C
Density: 0.95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00132754
CAS Number: 8012-89-3
EINECS: 232-383-7