Chloroform-[D₁] for NMR spectroscopy, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Trichloromethane-D1, Chloroform-[²H₁], Chloroform-D
M-Clarity™ quality level = MQ200
494275-10G 494275-50G
SIAL494275-10GEA 60.1 EUR
SIAL494275-10G SIAL494275-50G
Chloroform-[D₁] for NMR spectroscopy, Sigma-Aldrich®
Chloroform-[D1]
Formula: CDCl₃
Boiling Pt: 60.8 °C (1013 hPa)
Melting Pt: –64.1 °C
Density: 1.5 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000827
CAS Number: 865-49-6
EINECS: 212-742-4
UN: 1888
ADR: 6.1,III