Chloroform-[D₁] for NMR spectroscopy, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Trichloromethane-D1, Chloroform-[²H₁], Chloroform-D

M-Clarity™ quality level = MQ200

434876-100G 434876-150G 434876-500G
SIAL434876-100GEA 63.3 EUR
SIAL434876-100G SIAL434876-150G SIAL434876-500G
Chloroform-[D₁] for NMR spectroscopy, Sigma-Aldrich®
Chloroform-[D1]
Formula: CDCl₃
Boiling Pt: 60.8 °C (1013 hPa)
Melting Pt: –64.1 °C
Density: 1.5 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000827
CAS Number: 865-49-6
EINECS: 212-742-4
UN: 1888
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR